Methyl-4-iodobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19342-09
A19342-18
AAA19342-09EA
49.7
USD
AAA19342-09
AAA19342-18
Methyl-4-iodobenzoate 98%
Methyl-4-iodobenzoate
Purity: 98%
CAS No.: 619-44-3
Molecular Formula: C8H7IO2
Formula Weight: 262.05
Melting Point: 114-116°
MDL No.: MFCD00016353
Formula:
C₈H₇IO₂ Boiling Pt: 276 °C (1013 hPa) Melting Pt: 114…116 °C Density: 2.02 g/cm³ (10 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016353 CAS Number: 619-44-3 EINECS: 210-597-1 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...