5-Methylquinoxaline 98%

Supplier: Thermo Scientific Chemicals
Warning

A19300-06 A19300-03
AAA19300-06EA 79.9 USD
AAA19300-06 AAA19300-03
5-Methylquinoxaline 98%
5-Methylquinoxaline

Purity: 98% CAS No.: 13708-12-8 Molecular Formula: C9H8N2 Formula Weight: 144.18 Flash Point: 110°(230°F) Refractive Index: 1.6200 Melting Point: 20-21° Boiling Point: 119-121°/15mm MDL No.: MFCD00012335

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 119…121 °C/15 mm
Melting Pt: 20…21 °C
Density: 1,12
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00012335
CAS Number: 13708-12-8
EINECS: 237-246-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR