5-Methylcaproic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Methylhexanoic acid, 5-Methylhexanoicacid
A19012-09
A19012-18
AAA19012-09EA
78.25
USD
AAA19012-09
AAA19012-18
5-Methylcaproic acid 98%
5-Methylcaproic acid
Purity: 98%
CAS No.: 628-46-6
Molecular Formula: (CH3)2CH(CH2)3CO2H
Formula Weight: 130.19
Flash Point: 102°(216°F)
Refractive Index: 1.4220
Odor: Rancid
Boiling Point: 216°
UN No.: 3265
MDL No.: MFCD00036494
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Boiling Pt: 216 °C Density: 0.91 Flash Pt: 102 °C (216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036494 CAS Number: 628-46-6 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...