Dimethyl maleate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyl Maleate, Maleic acid dimethyl ester

A18944-0I A18944-22 A18944-0B A18944-30
AAA18944-0IEA 573 USD
AAA18944-0I AAAA18944-22 AAA18944-0B AAA18944-30
Dimethyl maleate 96%
Dimethyl maleate

Purity: 96% CAS No.: 624-48-6 Molecular Formula: C6H8O4 Formula Weight: 144.13 Flash Point: 91°(195°F) Refractive Index: 1.4420 Melting Point: -19° Boiling Point: 204-205° Beilstein Reference: 471705 UN No.: 2810 MDL No.: MFCD00008459

Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 204…205 °C
Melting Pt: –19 °C
Density: 1.153 g/cm³ (20 °C)
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008459
CAS Number: 624-48-6
EINECS: 210-848-5
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR