Calcium formate 98%

Supplier: Thermo Scientific Chemicals
Danger

A18916-22 A18916-36
AAA18916-22EA 57.02 USD
AAA18916-22 AAA18916-36
Calcium formate 98%
Calcium formate

Purity: 98% CAS No.: 544-17-2 Molecular Formula: Ca(O2CH)2 Formula Weight: 130.12 Melting Point: 300° MDL No.: MFCD00036108

Formula: C₂H₂CaO₄
Melting Pt: >300 °C
Density: 2.02
Storage Temperature: Ambient
MDL Number: MFCD00036108
CAS Number: 544-17-2
EINECS: 208-863-7
Merck Index: 12,01711

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR