(±)-3-Methylvaleric acid 99%

Supplier: Thermo Scientific Chemicals
Danger

A18886-14 A18886-22 A18886-06
AAA18886-14EA 117.73 USD
AAA18886-14 AAAA18886-22 AAA18886-06
(±)-3-Methylvaleric acid 99%
(±)-3-Methylvaleric acid

Purity: 99% CAS No.: 105-43-1 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 85°(185°F) Refractive Index: 1.4160 Odor: Unpleasant Melting Point: -42° Boiling Point: 196-198° Beilstein Reference: 1720696 UN No.: 3265 MDL No.: MFCD00002729

Formula: C₆H₁₂O₂
Boiling Pt: 196…198 °C
Melting Pt: –42 °C
Density: 0.930
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002729
CAS Number: 105-43-1
EINECS: 203-297-7
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR