N,N-Dimethylbenzamide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A18766-14 A18766-36 A18766-22
AAA18766-14EA 41.27 USD
AAA18766-14 AAAA18766-36 AAA18766-22
N,N-Dimethylbenzamide 98+%
N,N-Dimethylbenzamide

Purity: 98+% CAS No.: 611-74-5 Molecular Formula: C9H11NO Formula Weight: 149.19 Flash Point: 110°(230°F) Melting Point: 42-45° Boiling Point: 132-133°/15mm Beilstein Reference: 774955 MDL No.: MFCD00008320

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Boiling Pt: 132…133 °C/15 mm
Melting Pt: 42…45 °C
Density: 1.11 g/cm³ (23 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008320
CAS Number: 611-74-5
EINECS: 210-279-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR