4-Iodo-2-nitrotoluene 97%

Supplier: Thermo Scientific Chemicals
Warning

A18607-06 A18607-14
AAA18607-22EA 1509.15 USD
AAA18607-22 AAA18607-06 AAA18607-14
4-Iodo-2-nitrotoluene 97%
4-Iodo-2-nitrotoluene

Purity: 97% CAS No.: 41252-97-5 Molecular Formula: C7H6INO2 Formula Weight: 263.03 Melting Point: 58-62° MDL No.: MFCD00051090

Formula: C₇H₆INO₂
MW: 263.03 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00051090
CAS Number: 41252-97-5
EINECS: 255-279-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR