Gum tragacanth, powder
Supplier: Thermo Scientific Chemicals
A18502-36
A18502-22
AAA18502-36EA
223.57
USD
AAA18502-36
AAA18502-22
Gum tragacanth, powder
Gum tragacanth
CAS No.: 9000-65-1
Odor: Odorless
Merck Reference: 14,4581
MDL No.: MFCD00131255
Storage Temperature:
Ambient |
MDL Number:
MFCD00131255 CAS Number: 9000-65-1 EINECS: 232-552-5 Merck Index: 14,4581 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...