(Iodomethyl)trimethylammonium iodide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Iodomethylenetrimethylammonium iodide
A18424-14
A18424-06
AAA18424-14EA
129
USD
AAA18424-14
AAAA18424-22
AAA18424-06
(Iodomethyl)trimethylammonium iodide 98+%
(Iodomethyl)trimethylammonium iodide
Purity: 98+%
CAS No.: 39741-91-8
Molecular Formula: (CH3)3N(CH2I)I
Formula Weight: 326.94
Melting Point: ca 190° dec.
Beilstein Reference: 3912137
Formula:
C₄H₁₁I₂N MW: 326.95 g/mol Melting Pt: ∼ 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00031647 CAS Number: 39741-91-8 EINECS: 254-611-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...