Pentafluorobenzamide 99%

Supplier: Thermo Scientific Chemicals
Warning

A18397-06 A18397-14 A18397-22
AAA18397-06EA 74.3 USD
AAA18397-06 AAA18397-14 AAA18397-22
Pentafluorobenzamide 99%
Pentafluorobenzamide

Purity: 99% CAS No.: 652-31-3 Molecular Formula: C7H2F5NO Formula Weight: 211.09 Melting Point: 146-149° MDL No.: MFCD00007971

Formula: C₇H₂F₅NO
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00007971
CAS Number: 652-31-3
EINECS: 211-488-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR