2-Fluorophenyl isothiocyanate 97%

Supplier: Thermo Scientific Chemicals
Warning

A18314-06 A18314-14
AAA18314-06EA 44.9 USD
AAA18314-06 AAA18314-14
2-Fluorophenyl isothiocyanate 97%
2-Fluorophenyl isothiocyanate

Purity: 97% CAS No.: 38985-64-7 Molecular Formula: C7H4FNS Formula Weight: 153.18 Flash Point: 97°(206°F) Refractive Index: 1.6254 Boiling Point: 103-104°/12mm Beilstein Reference: 4176581 UN No.: 2922 MDL No.: MFCD00004800

Formula: C₇H₄FNS
MW: 153.18 g/mol
Boiling Pt: 103…104 °C/12 mm
Melting Pt: 24 - 26 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 97 °C (206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004800
CAS Number: 38985-64-7
EINECS: 254-237-1
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR