4-Phenylbutyric acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Phenylbutanoic acid

A18115-14 A18115-22 A18115-36
AAA18115-14EA 65.38 USD
AAA18115-14 AAA18115-22 AAAA18115-36
4-Phenylbutyric acid 99%
4-Phenylbutyric acid

Purity: 99% CAS No.: 1821-12-1 Molecular Formula: C6H5(CH2)3CO2H Formula Weight: 164.20 Flash Point: 110°(230°F) Melting Point: 49-53° Boiling Point: 165°/10mm Beilstein Reference: 638180 MDL No.: MFCD00004403

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 165 °C/10 mm
Melting Pt: 49…53 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004403
CAS Number: 1821-12-1
EINECS: 217-341-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR