2-(Cyclohexylamino)ethanesulphonic acid (CHES) 99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Cyclohexyltaurine, CHES

A18047-14 A18047-36 A18047-22
AAA18047-14EA 82.78 USD
AAA18047-14 AAAA18047-36 AAA18047-22
2-(Cyclohexylamino)ethanesulphonic acid (CHES) 99%
2-(Cyclohexylamino)ethanesulphonic acid (CHES)

Zwitterionic buffer useful in pH range 8.6 to 10.0


Purity: 99% CAS No.: 103-47-9 Molecular Formula: C8H17NO3S Formula Weight: 207.29 Melting Point: 300° Beilstein Reference: 2967601 Merck Reference: 14,2055 MDL No.: MFCD00003835


Zwitterionic buffer useful in pH range 8.6 to 10.0

Formula: C₈H₁₇NO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003835
CAS Number: 103-47-9
EINECS: 203-115-6
Merck Index: 13,02063

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR