6-Methyl-2-thiouracil 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methiocil, 2-Mercapto-6-methylpyrimidin-4(1H)-one, Methylthiouracil, MZU, Basethyrin, 4-Hydroxy-2-mercapto 6-methylpyrimidine, Thiothymin

A17982-18 A17982-30 A17982-A1
AAA17982-18EA 24.7 USD
AAA17982-18 AAA17982-30 AAAA17982-A1
6-Methyl-2-thiouracil 98%
6-Methyl-2-thiouracil

Purity: 98% CAS No.: 56-04-2 Molecular Formula: C5H6N2OS Formula Weight: 142.18 Melting Point: ca 329° dec. Beilstein Reference: 115648 Merck Reference: 14,6128 MDL No.: MFCD00006040

Formula: C₅H₆N₂OS
MW: 142.18 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: ∼ 329 °C (dec.)
Density: 1.519 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006040
CAS Number: 56-04-2
EINECS: 200-252-3
Merck Index: 12,06210

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR