Triethylammonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Triethylamine hydrobromide

A17926-36 A17926-22
AAA17926-36EA 155 USD
AAA17926-36 AAA17926-22
Triethylammonium bromide 98%
Triethylammonium bromide

Triethylamine hydrobromide is used to prepare dimethyl -alfa-diethylaminomaleate by reacting with dimethyl acetylenedicarbosylate. It is involved in the preparation of triphenylpropargylphosphonium bromide.


Purity: 98% CAS No.: 636-70-4 Molecular Formula: C16H15N.HBr Formula Weight: 182.10 Odor: Amine-like Melting Point: ca 250° dec. MDL No.: MFCD00054287

Formula: C₆H₁₅N.HBr
MW: 182.1 g/mol
Melting Pt: ca 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00054287
CAS Number: 636-70-4
EINECS: 211-263-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR