Tetramethylphosphonium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17907-06
A17907-03
AAA17907-06EA
226
USD
AAA17907-06
AAA17907-03
Tetramethylphosphonium bromide 98%
Tetramethylphosphonium bromide
Purity: 98%
CAS No.: 4519-28-2
Molecular Formula: (CH3)4PBr
Formula Weight: 171.03
MDL No.: MFCD00011802
Formula:
C₄H₁₂BrP Storage Temperature: Ambient |
MDL Number:
MFCD00011802 CAS Number: 4519-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...