Cyclooctanol 97%

Supplier: Thermo Scientific Chemicals
Warning

A17825-14 A17825-22
AAA17825-14EA 76 USD
AAA17825-14 AAA17825-22
Cyclooctanol 97%
Cyclooctanol

Purity: 97% CAS No.: 696-71-9 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 86°(186°F) Refractive Index: 1.4860 Melting Point: 14-15° Boiling Point: 97-98°/11mm Beilstein Reference: 1848166 MDL No.: MFCD00001744

Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 97…98 °C/11 mm
Melting Pt: 14…15 °C
Density: 0.97 g/cm³ (20 °C)
Flash Pt: 86 °C(186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001744
CAS Number: 696-71-9
EINECS: 211-800-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR