Sodium L(+)-ascorbate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt

A17759-36 A17759-22
AAA17759-36EA 173.41 USD
AAA17759-36 AAA17759-22
Sodium L(+)-ascorbate 99%
Sodium L(+)-ascorbate

Purity: 99% CAS No.: 134-03-2 Molecular Formula: C6H7NaO6 Formula Weight: 198.11 Melting Point: ca 220° Beilstein Reference: 3767246 Merck Reference: 14,830 Optical Rotation: +105° (c=5 in water) MDL No.: MFCD00082340

Formula: C₆H₇NaO₆
MW: 198.11 g/mol
Melting Pt: ∼ 220 °C
Density: 1.799 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082340
CAS Number: 134-03-2
EINECS: 205-126-1
Merck Index: 14,830
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d142b281353c28f2a72502b60fece03e926023d&vid=988032859afe97984b0fc1b8bfd43cff093f099a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR