2-Pentene cis- and trans-mixture 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17736-14
A17736-22
AAA17736-14EA
97.54
USD
AAA17736-14
AAA17736-22
2-Pentene cis- and trans-mixture 98%
2-Pentene
Purity: 98%
CAS No.: 109-68-2
Molecular Formula: C5H10
Formula Weight: 70.14
Storage Temperature: 2°C to 8°C
Flash Point: -45°(-49°F)
Refractive Index: 1.38
Odor: Petroleum-like
Boiling Point: 36-38°
Beilstein Reference: 969151
Merck Reference: 14,7123
UN No.: 3295
MDL No.: MFCD00009384
Formula:
C₅H₁₀ MW: 70.13 g/mol Boiling Pt: 36…38 °C Melting Pt: -140 °C Density: 0.658 g/cm³ (20 °C) Flash Pt: –45 °C (–49 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009384 CAS Number: 109-68-2 EINECS: 203-695-0 UN: 3295 ADR: 3,III Merck Index: 12,07263 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...