2,2'-Dibromobiphenyl (PBB No. 4) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-2-(2-bromophenyl)benzene, 2,2'-Dibromobiphenyl

A17632-14 A17632-03 A17632-06
AAA17632-14EA 754 USD
AAA17632-14 AAA17632-03 AAA17632-06
2,2'-Dibromobiphenyl (PBB No. 4) 98%
2,2'-Dibromobiphenyl (PBB No. 4)

Purity: 98% CAS No.: 13029-09-9 Molecular Formula: C12H8Br2 Formula Weight: 312.02 Melting Point: 78-81° UN No.: 3152 MDL No.: MFCD00093707

Formula: C₁₂H₈Br₂
MW: 312 g/mol
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00093707
CAS Number: 13029-09-9
UN: 3152
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR