Acetoacetanilide 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Oxo-N-phenyl butamide, 3-Oxo-N-phenyl butanamide

A17534-0B A17534-30
AAA17534-0BEA 53 USD
AAA17534-0B AAA17534-0I AAA17534-30
Acetoacetanilide 98+%
Acetoacetanilide

Purity: 98+% CAS No.: 102-01-2 Molecular Formula: C6H5NHCOCH2COCH3 Formula Weight: 177.20 Flash Point: 150°(302°F) Odor: Odorless Boiling Point: 129°/24mm Beilstein Reference: 473419 Merck Reference: 14,58 MDL No.: MFCD00008780

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Boiling Pt: 129 °C/24 mm
Melting Pt: 83…86 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 150 °C(302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008780
CAS Number: 102-01-2
EINECS: 202-996-4
Merck Index: 13,00059

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR