Bilirubin 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-(2-((3-(2-Carboxyethyl)-4-methyl-5-((Z)-(3-methyl-5-oxo-4-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-2-yl)methyl)-4-methyl-5-((Z)-(4-methyl-5-oxo-3-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-3-yl)propanoic acid

A17522-03 A17522-14 A17522-06
AAA17522-03EA 288.92 USD
AAA17522-03 AAA17522-14 AAA17522-06
Bilirubin 97%
Bilirubin

Purity: 97% CAS No.: 635-65-4 Molecular Formula: C33H36N4O6 Formula Weight: 584.68 Beilstein Reference: 74376 Merck Reference: 14,1218 MDL No.: MFCD00005499

Formula: C₃₃H₃₆N₄O₆
MW: 584.67 g/mol
Melting Pt: > 360 °C
Storage Temperature: Ambient
MDL Number: MFCD00005499
CAS Number: 635-65-4
EINECS: 211-239-7
Merck Index: 14,1218

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR