3-Phenoxyaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Amino-3-phenoxybenzol

A17495-14 A17495-06 A17495-22
AAA17495-14EA 138 USD
AAA17495-14 AAA17495-06 AAA17495-22
3-Phenoxyaniline 98%
3-Phenoxyaniline

Purity: 98% CAS No.: 3586-12-7 Molecular Formula: C12H11NO Formula Weight: 185.23 Flash Point: 110°(230°F) Refractive Index: 1.1583 Melting Point: 35-37° Boiling Point: 329-330° MDL No.: MFCD00041891

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Boiling Pt: 329…330 °C
Melting Pt: 35…37 °C
Density: 1.1584 g/cm³ (25 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041891
CAS Number: 3586-12-7
EINECS: 222-715-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR