4-Chlorobenzophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Chlorobenzophenone

A17458-36 A17458-0E A17458-22
AAA17458-36EA 117 USD
AAA17458-36 AAA17458-0E AAA17458-22
4-Chlorobenzophenone 99%
4-Chlorobenzophenone

Purity: 99% CAS No.: 134-85-0 Molecular Formula: C13H9ClO Formula Weight: 216.67 Flash Point: 143°(289°F) Melting Point: 75-77° Boiling Point: 332° Beilstein Reference: 512043 MDL No.: MFCD00000622

Formula: C₁₃H₉ClO
MW: 216.67 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 68 - 70 °C
Flash Pt: 143 °C(289 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000622
CAS Number: 134-85-0
EINECS: 205-160-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR