Tetramethylammonium tetrafluoroborate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetramethylammoniumtetrafluoroborate

A17436-14 A17436-22 A17436-06
AAA17436-14EA 82.12 USD
AAA17436-14 AAA17436-22 AAA17436-06
Tetramethylammonium tetrafluoroborate 97%
Tetramethylammonium tetrafluoroborate

Purity: 97% CAS No.: 661-36-9 Molecular Formula: (CH3)4NBF4 Formula Weight: 160.95 Melting Point: 417-419° Beilstein Reference: 3715404

Formula: C₄H₁₂BF₄N
Melting Pt: 418 °C
Storage Temperature: Ambient
MDL Number: MFCD00011745
CAS Number: 661-36-9
EINECS: 211-547-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR