3-Chloroanisole 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Chloro-3-methoxybenzene

A17327-30 A17327-18
AAA17327-30EA 236 USD
AAA17327-30 AAA17327-18
3-Chloroanisole 98+%
3-Chloroanisole

Purity: 98+% CAS No.: 2845-89-8 Molecular Formula: C7H7ClO Formula Weight: 142.59 Flash Point: 73°(163°F) Refractive Index: 1.5360 Boiling Point: 192-194° Beilstein Reference: 2041497 MDL No.: MFCD00000591

Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 192…194 °C
Density: 1.172 g/cm³ (20 °C)
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000591
CAS Number: 2845-89-8
EINECS: 220-642-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR