Tetrabutylammonium hexafluorophosphate 98%

Supplier: Thermo Scientific Chemicals
Warning

A17196-14 A17196-22 A17196-36
AAA17196-14EA 93.49 USD
AAA17196-14 AAA17196-22 AAA17196-36
Tetrabutylammonium hexafluorophosphate 98%
Tetrabutylammonium hexafluorophosphate

Purity: 98% CAS No.: 3109-63-5 Molecular Formula: C16H36F6NP Formula Weight: 387.44 Melting Point: 242-248° MDL No.: MFCD00011748

Formula: C₁₆H₃₆F₆NP
MW: 387.43 g/mol
Melting Pt: 242…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00011748
CAS Number: 3109-63-5
EINECS: 221-472-6
UN: 3263
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR