(±)-1,2-Dibromohexafluoropropane 95%

Supplier: Thermo Scientific Chemicals

A17103-14 A17103-22
AAA17103-14EA 43.9 USD
AAA17103-14 AAA17103-22
(±)-1,2-Dibromohexafluoropropane 95%
(±)-1,2-Dibromohexafluoropropane

Purity: 95% CAS No.: 661-95-0 Molecular Formula: BrCF2CFBrCF3 Formula Weight: 309.84 Boiling Point: 71-72° MDL No.: MFCD00013568

Formula: BrF₂CBrCFCF₃
Boiling Pt: 71…72 °C
Density: 2.169
Storage Temperature: Ambient
MDL Number: MFCD00013568
CAS Number: 661-95-0
EINECS: 211-550-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR