4-Aminophthalimide 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
5-Aminoisoindoline-1,3-dione, 5-Amino-isoindole-1,3-dione
A17051-14
A17051-06
AAA17051-14EA
331
USD
AAA17051-14
AAA17051-06
4-Aminophthalimide 97%
4-Aminophthalimide
Purity: 97%
CAS No.: 3676-85-5
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
Melting Point: 289-291°
MDL No.: MFCD00041854
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol Boiling Pt: 447 Melting Pt: 294 Storage Temperature: Ambient |
MDL Number:
MFCD00041854 CAS Number: 3676-85-5 EINECS: 222-948-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...