1,2,3-Triazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,2,3-1H-Triazole, 1H-1,2,3-Triazole
A16999-03
A16999-06
A16999-14
AAA16999-03EA
55.8
USD
AAA16999-03
AAA16999-06
AAAA16999-14
1,2,3-Triazole 98%
1,2,3-Triazole
Purity: 98%
CAS No.: 288-36-8
Molecular Formula: C2H3N3
Formula Weight: 69.07
Flash Point: 107°(224°F)
Refractive Index: 1.4980
Melting Point: 23-25°
Boiling Point: 208°
Beilstein Reference: 104766
MDL No.: MFCD00014490
Formula:
C₂H₃N₃ Boiling Pt: 208 °C Melting Pt: 23…25 °C Density: 1.192 g/cm³ (20 °C) Flash Pt: 107 °C(224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014490 CAS Number: 288-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...