1,3-Dibutyl-2-thiourea 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N,N'-Dibutylthiourea, 1,3-Dibutylthiourea, N,N'-Di-n-butylthiourea
A16783-0B
A16783-30
A16783-18
AAA16783-0BEA
178
USD
AAA16783-0B
AAA16783-30
AAA16783-18
1,3-Dibutyl-2-thiourea 98%
1,3-Dibutyl-2-thiourea
Purity: 98%
CAS No.: 109-46-6
Molecular Formula: CH3(CH2)3NHCSNH(CH2)3CH3
Formula Weight: 188.34
Odor: Odorless
Melting Point: 64-67°
Beilstein Reference: 507434
MDL No.: MFCD00004926
Formula:
H₃CCH₂CH₂CH₂NHC(S)NHCH₂CH₂CH₂CH₃ MW: 188.34 g/mol Boiling Pt: 252 Melting Pt: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004926 CAS Number: 109-46-6 EINECS: 203-674-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...