3-Benzoylpropionic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Oxo-4-phenylbutanoic acid, 3-Benzoylpropionicacid
A16636-14
A16636-22
AAA16636-14EA
59.7
USD
AAA16636-14
AAA16636-22
3-Benzoylpropionic acid 98+%
3-Benzoylpropionic acid
Purity: 98+%
CAS No.: 2051-95-8
Molecular Formula: C6H5COCH2CH2CO2H
Formula Weight: 178.19
Melting Point: 114-120°
Beilstein Reference: 639757
Formula:
C₁₀H₁₀O₃ MW: 178.19 g/mol Boiling Pt: 325 °C (1013 hPa) Melting Pt: 114 - 117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002792 CAS Number: 2051-95-8 EINECS: 218-135-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...