N,N-Dimethyl-1,1,1-trimethylsilylamine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (Dimethylamino)trimethylsilane, Trimethylsilyldimethylamine, N,N-Dimethyltrimethylsilylamine, TMSDMA, N-(Trimethylsilyl)dimethylamine, N,N-Dimethylaminotrimethylsilane, Pentamethylsilylamine

A16550-09 A16550-18
AAA16550-09EA 42.5 USD
AAA16550-09 AAA16550-18
N,N-Dimethyl-1,1,1-trimethylsilylamine 95%
N,N-Dimethyl-1,1,1-trimethylsilylamine

Purity: 95% CAS No.: 2083-91-2 Molecular Formula: C5H15SiN Formula Weight: 117.27 Flash Point: -19°(-3°F) Refractive Index: 1.3970 Odor: Amine-like Boiling Point: 86-87° Beilstein Reference: 1731861 UN No.: 2733 MDL No.: MFCD00008297

Formula: C₅H₁₅NSi
MW: 117.27 g/mol
Boiling Pt: 84 °C (1013 hPa)
Density: 0.7466 g/cm³ (20 °C)
Flash Pt: –19 °C (–3 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008297
CAS Number: 2083-91-2
EINECS: 218-222-3
UN: 2733
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR