3,5-Difluoroaniline 98+%

Supplier: Thermo Scientific Chemicals
Warning

A16528-06 A16528-14
AAAA16528-22EA 1083.63 USD
AAAA16528-22 AAA16528-06 AAAA16528-14
3,5-Difluoroaniline 98+%
3,5-Difluoroaniline

Purity: 98+% CAS No.: 372-39-4 Molecular Formula: C6H5F2N Formula Weight: 129.11 Flash Point: 75°(167°F) Odor: Characteristic Boiling Point: 80°/20mm Beilstein Reference: 2802286 UN No.: 2811 MDL No.: MFCD00007763

Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 80 °C/20 mm
Melting Pt: 37…41 °C
Density: 1.295 g/cm³ (20 °C)
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007763
CAS Number: 372-39-4
EINECS: 206-752-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR