Cyclohexylbenzene 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenylcyclohexane

A16320-22 A16320-36
AAA16320-22EA 59.2 USD
AAA16320-22 AAA16320-36
Cyclohexylbenzene 97+%
Cyclohexylbenzene

Purity: 97+% CAS No.: 827-52-1 Molecular Formula: C12H16 Formula Weight: 160.26 Flash Point: 98°(208°F) Refractive Index: 1.5260 Odor: Mild Melting Point: 5-6° Boiling Point: 239-240° Beilstein Reference: 1906803 UN No.: 3082 MDL No.: MFCD00001451

Formula: C₁₂H₁₆
MW: 160.26 g/mol
Boiling Pt: 239…240 °C
Melting Pt: 5…6 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001451
CAS Number: 827-52-1
EINECS: 212-572-0
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR