Dibutyl ether 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di-n-butyl ether

A16305-AP A16305-0F
AAA16305-APEA 33.47 USD
AAA16305-AP AAA16305-0F
Dibutyl ether 99%
Dibutyl ether

Purity: 99% CAS No.: 142-96-1 Molecular Formula: CH3(CH2)3O(CH2)3CH3 Formula Weight: 130.23 Flash Point: 25°(77°F) Refractive Index: 1.3990 Melting Point: -95° Boiling Point: 141-143° Beilstein Reference: 1732752 Merck Reference: 14,1569 UN No.: 1149 MDL No.: MFCD00009461

Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 141…143 °C
Melting Pt: –95 °C
Density: 0.767 g/cm³ (20 °C)
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009461
CAS Number: 142-96-1
EINECS: 205-575-3
UN: 1149
ADR: 3,III
Merck Index: 13,01568

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR