1,4-Dichlorobenzene 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Dichlorobenzene

A16290-36 A16290-22 A16290-0E
AAA16290-36EA 30.23 USD
AAA16290-36 AAAA16290-22 AAA16290-0E
1,4-Dichlorobenzene 99+%
1,4-Dichlorobenzene

Purity: 99+% CAS No.: 106-46-7 Molecular Formula: C6H4Cl2 Formula Weight: 147.00 Flash Point: 65°(149°F) Odor: Strong Boiling Point: 173-174° Beilstein Reference: 1680023 Merck Reference: 14,3057 UN No.: 3077 MDL No.: MFCD00000604

Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 173…174 °C
Melting Pt: 52…56 °C
Density: 1.458 g/cm³ (20 °C)
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000604
CAS Number: 106-46-7
EINECS: 203-400-5
UN: 3077
ADR: "9",III
Merck Index: 13,03082

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR