D-(-)-Arginine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
A16222-03
A16222-06
A16222-14
AAA16222-03EA
42.4
USD
AAA16222-03
AAA16222-06
AAA16222-14
D-(-)-Arginine hydrochloride 99%
D-(-)-Arginine hydrochloride
Purity: 99%
CAS No.: 627-75-8
Molecular Formula: C6H14N4O2.HCl
Formula Weight: 210.67
Melting Point: 216-218°
Beilstein Reference: 5773432
Formula:
C₆H₁₅ClN₄O₂ Melting Pt: 216…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012620 CAS Number: 627-75-8 EINECS: 211-010-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...