Undecanal 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Hendecanal, Undecylaldehyde
A16101-22
A16101-14
AAA16101-22EA
99.5
USD
AAA16101-22
AAA16101-14
Undecanal 97%
Undecanal
Purity: 97%
CAS No.: 112-44-7
Molecular Formula: C11H22O
Formula Weight: 170.30
Flash Point: 95°(203°F)
Refractive Index: 1.4340
Melting Point: -2°
Boiling Point: 110-112°/12mm
Beilstein Reference: 1753213
MDL No.: MFCD00007016
Formula:
C₁₁H₂₂O MW: 170.3 g/mol Boiling Pt: 226 °C (1013 hPa) Melting Pt: -2 °C Density: 0.827 g/cm³ (20 °C) Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007016 CAS Number: 112-44-7 EINECS: 203-972-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...