4,7-Dichloroquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

A15985-36 A15985-14 A15985-22
AAAA15985-36EA 968 USD
AAAA15985-36 AAA15985-14 AAA15985-22
4,7-Dichloroquinoline 98%
4,7-Dichloroquinoline

Purity: 98% CAS No.: 86-98-6 Molecular Formula: C9H5Cl2N Formula Weight: 198.05 Flash Point: 164°(327°F) Melting Point: 83-87° Boiling Point: 148°/10mm Beilstein Reference: 125359 MDL No.: MFCD00006774

Formula: C₉H₅Cl₂N
MW: 198.05 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 81 - 83 °C
Flash Pt: 164 °C(327 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006774
CAS Number: 86-98-6
EINECS: 201-714-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR