4-Methoxybenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile

A15945-14 A15945-06 A15945-22
AAA15945-14EA 106 USD
AAA15945-14 AAA15945-06 AAA15945-22
4-Methoxybenzonitrile 99%
4-Methoxybenzonitrile

Purity: 99% CAS No.: 874-90-8 Molecular Formula: C8H7NO Formula Weight: 133.15 Odor: Almond-like Melting Point: 57-61° Boiling Point: 256-257° Beilstein Reference: 508117 UN No.: 3439 MDL No.: MFCD00001818

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 256…257 °C
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00001818
CAS Number: 874-90-8
EINECS: 212-871-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR