3-Cyanobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Formylbenzonitrile
A15943-03
A15943-06
A15943-14
AAA15943-03EA
20.3
USD
AAA15943-03
AAA15943-06
AAA15943-14
3-Cyanobenzaldehyde 97%
3-Cyanobenzaldehyde
Purity: 97%
CAS No.: 24964-64-5
Molecular Formula: C8H5NO
Formula Weight: 131.13
Odor: Characteristic
Melting Point: 75-79°
Boiling Point: 209-210°
Beilstein Reference: 2205751
MDL No.: MFCD00003344
Formula:
C₈H₅NO MW: 131.13 g/mol Boiling Pt: 209…210 °C Melting Pt: 75…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003344 CAS Number: 24964-64-5 EINECS: 246-549-1 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...