Triphenylamine 98%

Supplier: Thermo Scientific Chemicals
Warning

A15939-14 A15939-22 A15939-06
AAA15939-14EA 64.88 USD
AAA15939-14 AAA15939-22 AAA15939-06
Triphenylamine 98%
Triphenylamine

Purity: 98% CAS No.: 603-34-9 Molecular Formula: (C6H5)3N Formula Weight: 245.33 Melting Point: 125-129° Boiling Point: 365° MDL No.: MFCD00003020

Formula: C₁₈H₁₅N
MW: 245.32 g/mol
Boiling Pt: 365 °C
Melting Pt: 125…129 °C
Density: 0.774 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003020
CAS Number: 603-34-9
EINECS: 210-035-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR