(3,4-Dihydroxyphenyl)acetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DOPAC, 3,4-Dihydroxybenzeneacetic acid, 3,4-Dihydroxyphenylacetic acid

A15893-06 A15893-14 A15893-03
AAA15893-06EA 85 USD
AAA15893-06 AAA15893-14 AAA15893-03
(3,4-Dihydroxyphenyl)acetic acid 98+%
(3,4-Dihydroxyphenyl)acetic acid

Purity: 98+% CAS No.: 102-32-9 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 128-130° Beilstein Reference: 2211017

Formula: (C₆H₃(OH)₂)CH₂COOH
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004338
CAS Number: 102-32-9
EINECS: 203-024-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR