Tetraphenylphosphonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15860-18 A15860-09 A15860-22
AAA15860-18EA 120 USD
AAA15860-18 AAA15860-09 AAAA15860-22
Tetraphenylphosphonium bromide 98+%
Tetraphenylphosphonium bromide

Purity: 98+% CAS No.: 2751-90-8 Molecular Formula: (C6H5)4PBr Formula Weight: 419.31 Melting Point: 296-302° Beilstein Reference: 3922383 Merck Reference: 14,9237 MDL No.: MFCD00011915

Formula: C₂₄H₂₀BrP
MW: 419.3 g/mol
Melting Pt: 296…302 °C
Storage Temperature: Ambient
MDL Number: MFCD00011915
CAS Number: 2751-90-8
EINECS: 220-393-4
Merck Index: 14,9237

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR