Cyclohexylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aminocyclohexane

A15851-AP A15851-AE A15851-0F
AAA15851-APEA 55.86 USD
AAA15851-AP AAAA15851-AE AAA15851-0F
Cyclohexylamine 98+%
Cyclohexylamine

Purity: 98+% CAS No.: 108-91-8 Molecular Formula: C6H13N Formula Weight: 99.18 Flash Point: 27°(81°F) Refractive Index: 1.4585 Melting Point: -18° Boiling Point: 133-134° Beilstein Reference: 471175 Merck Reference: 14,2729 UN No.: 2357 MDL No.: MFCD00001486

Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 134.5 °C (1013 hPa)
Melting Pt: -18 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00001486
CAS Number: 108-91-8
EINECS: 203-629-0
UN: 2357
ADR: 8,II
Merck Index: 13,02758

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR