Methyl formate 97% may cont. up to ca 3% methanol

Supplier: Thermo Scientific Chemicals
Danger

A15850-0F A15850-0D A15850-AP
AAA15850-0FEA 96.14 USD
AAA15850-0F AAA15850-0D AAA15850-AP
Methyl formate 97% may cont. up to ca 3% methanol
Methyl formate

Purity: 97% CAS No.: 107-31-3 Molecular Formula: C2H4O2 Formula Weight: 60.05 Storage Temperature: 2°C to 8°C Flash Point: -26°(-14°F) Physical Form: Clear liquid Density: 0.972 Refractive Index: 1.3440 Appearance: Colorless Melting Point: -100° Boiling Point: 32-33° Beilstein Reference: 1734623 Merck Reference: 14,6077 UN No.: 1243 MDL No.: MFCD00003291

Formula: C₂H₄O₂
Boiling Pt: 32 °C (1013 hPa)
Melting Pt: -100.4 °C
Density: 0.9713 g/cm³ (20 °C)
Flash Pt: -32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003291
CAS Number: 107-31-3
EINECS: 203-481-7
UN: 1243
ADR: 3,I
Merck Index: 12,06153

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR