4-Bromotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15843-22
A15843-14
A15843-36
AAA15843-22EA
57.3
USD
AAA15843-22
AAAA15843-14
AAA15843-36
4-Bromotoluene 98%
4-Bromotoluene
Purity: 98%
CAS No.: 106-38-7
Molecular Formula: C7H7Br
Formula Weight: 171.04
Flash Point: 85°(185°F)
Melting Point: 26-29°
Boiling Point: 183-184°
Beilstein Reference: 1903636
Merck Reference: 14,1439
MDL No.: MFCD00000109
Formula:
C₇H₇Br Boiling Pt: 183…184 °C Melting Pt: 26…29 °C Density: 1.410 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000109 CAS Number: 106-38-7 EINECS: 203-391-8 UN: 3077 ADR: 9,III Merck Index: 14,1439 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...