6-Bromocaproic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A15761-22 A15761-36 A15761-14
AAA15761-22EA 192 USD
AAA15761-22 AAA15761-36 AAA15761-14
6-Bromocaproic acid 98+%
6-Bromocaproic acid

Purity: 98+% CAS No.: 4224-70-8 Molecular Formula: Br(CH2)5CO2H Formula Weight: 195.06 Flash Point: 95°(203°F) Odor: Odorless Boiling Point: 165-170°/20mm Beilstein Reference: 1749740 UN No.: 3261 MDL No.: MFCD00004422

Formula: BrH₂C(CH₂)₄COOH
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: 31 - 34 °C
Density: 1.4068 g/cm³ (30 °C)
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004422
CAS Number: 4224-70-8
EINECS: 224-176-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR